Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Silberchlorid ≥95%

Silberchlorid ≥95%

25 g
Catalog # 87342.14
Corrosive
Silberchlorid ≥95%
Catalog # 87342.14
CAS Number:  7783-90-6
Silberchlorid ≥95%
Silberchlorid ≥95%

25 g

CAS Number:  7783-90-6
Price
SFr. 256.00
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 95% CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Physical Form: Coarse Powder Density: 5.56 Refractive Index: 2.071 Sensitivity: Light Sensitive Appearance: White Odour: Odourless Melting Point: 455° Boiling Point: 1550° Solubility: Practically insoluble in water. Soluble in ammonia, solutions of alkali cyanides, thiosulfates, ammonium carbonates. Insoluble in alcohol, dilute acids Merck Reference: 14,8509 UN No.: 3077 MDL No.: MFCD00003399

Silver plating

  • CAS Number:7783-90-6
  • Formula:AgCl
  • MDL Number:MFCD00003399
  • Molecular Weight:143.32 g/mol
  • Merck Index:14,8509
  • Boiling Pt:1550 °C
  • Melting Pt:455 °C
  • Density:5,56
  • Storage Temperature:Raumtemperatur
  • EINECS:232-033-3
  • ADR:9,III
Specifications

Specifications

  • VE:
    25 g

Specifications