Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Benzalkoniumchlorid

Benzalkoniumchlorid

Glass bottle for solids, 500 g
Catalog # 41339.36
Corrosive
Irritant
Environment
Benzalkoniumchlorid
Catalog # 41339.36
CAS Number:  63449-41-2
Benzalkoniumchlorid
Benzalkoniumchlorid

Glass bottle for solids, 500 g

CAS Number:  63449-41-2
Price
441.00 SFr.
Log in for availability
1 * 500 g
Product Details & Documents

CAS No.: 63449-41-2 Molecular Formula: C6H5CH2N(CH3)2RCl Physical Form: Semi-solid/solid, R=C8H17 to C18H37 Solubility: Soluble in water Merck Reference: 14,1059 UN No.: 2923 MDL No.: MFCD00145757

Cationic surface active agent

Synonyms:  Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
  • CAS Number:63449-41-2
  • Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
  • MDL Number:MFCD00145757
  • Merck Index:14,1059
  • Melting Pt:29…34 °C
  • Density:0,98 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:264-151-6
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    500 g

Specifications