Benzalkoniumchlorid
Glass bottle for solids, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
CAS No.: 63449-41-2 Molecular Formula: C6H5CH2N(CH3)2RCl Physical Form: Semi-solid/solid, R=C8H17 to C18H37 Solubility: Soluble in water Merck Reference: 14,1059 UN No.: 2923 MDL No.: MFCD00145757
Cationic surface active agent
Synonyme: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
- CAS Nummer:63449-41-2
- Formel:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
- MDL-Nummer:MFCD00145757
- Merck-Index:14,1059
- Schmelzender Platingehalt:29…34 °C
- Dichte:0,98 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:264-151-6
- UN:2923
- ADR:8,III
Dokumente
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



