Natriumdihydrogenphosphat ≥96%
Glass bottle for solids, 250 g
Anbieter: Thermo Fisher Scientific
Natriumdihydrogenphosphat ≥96%
Katalognr. 40145.30
Anbieter: Thermo Fisher Scientific
CAS Nummer: 7558-80-7
Direkt zu
Product Details & Documents
Purity: 96% CAS No.: 7558-80-7 Molecular Formula: H2NaO4P Formula Weight: 119.98 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Solubility: Freely soluble in water. Insoluble in alcohol Merck Reference: 14,8660 MDL No.: MFCD00003527
Synonyme: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
- CAS Nummer:7558-80-7
- Formel:NaH₂PO₄
- MDL-Nummer:MFCD00003527
- Molekulargewicht:119.98 g/mol
- Merck-Index:14,8660
- Schmelzender Platingehalt:∼650 °C
- Dichte:0,9996 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:231-449-2
- ERREICHEN:01-2119489796-13
Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



