Infrared optical windows
Crystal optic disc, polished both sides, Zinc selenide, 13 mm, 2 mm
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
These optical crystal windows are ideal for infrared spectroscopy.
- Disc or rectangular
- Polished or unpolished
A polishing kit is available for those who wish to polish their windows.
Technische Daten
- Fenstermaterial:Zinc selenide
- Bezeichnung:Crystal optic disc, polished both sides
- Abmessungen:13 mm
- Seq:97
- Stärke:2 mm
Accessories
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis



