Essigsäure (Eisessig) ≥99%
Glasflasche, 20 L
Anbieter: Thermo Fisher Scientific
Essigsäure (Eisessig) ≥99%
Katalognr. 33252.M4
Anbieter: Thermo Fisher Scientific
CAS Nummer: 64-19-7
Direkt zu
Product Details & Documents
Purity: 99+% CAS No.: 64-19-7 Molecular Formula: CH3CO2H Formula Weight: 60.05 Flash Point: 40°(104°F) Physical Form: Liquid Density: 1.049 Refractive Index: 1.3721 Odour: Pungent pH: 2,5 Melting Point: 16.6° Boiling Point: 118.1° Merck Reference: 14,55 UN No.: 2789 MDL No.: MFCD00036152
Synonyme: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Nummer:64-19-7
- Formel:H₃CCOOH
- MDL-Nummer:MFCD00036152
- Molekulargewicht:60.05 g/mol
- Merck-Index:14,55
- Siedepunkt:118,1 °C
- Schmelzender Platingehalt:16,6 °C
- Dichte:1,049
- Blitz Pt:40 °C (104 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:200-580-7
- UN:2789
- ADR:8,II
Technische Daten
Technische Daten
- Packung:Glasflasche
- VE:20 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



