Silber(II)oxid ≥98%
5 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 1301-96-8 Molecular Formula: AgO Formula Weight: 123.87 Physical Form: Powder Density: 7.44 Appearance: Grey to black Odour: Odourless Melting Point: 100° dec. Solubility: Insoluble in water. Soluble in alkalies, ammonium hydroxide with decomposition and N2 formation Merck Reference: 14,8522 UN No.: 3085 MDL No.: MFCD00044285
Manufacturing of silver oxide-zinc alkali batteries
- CAS Nummer:1301-96-8
- Formel:AgO
- MDL-Nummer:MFCD00044285
- Molekulargewicht:123.87 g/mol
- Merck-Index:14,8522
- Schmelzender Platingehalt:>100 °C (Zers.)
- Schmelzender Platingehalt:>100 °C (Zersetzung)
- Dichte:7,44
- Lagertemperatur:Raumtemperatur
- EINECS:215-098-2
- UN:3085
- ADR:5.1,II
- :Density: 7,44 g/cm³ (20 °C)
Dokumente
Zertifikate
Suche nach Chargennummermsds_ALFA022695
Sicherheitsdatenblatt (SDS)
Thermo Scientific Silver Products
Produktliteratur
Technische Daten
Technische Daten
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



