Thymolblau ACS
Glass bottle for solids, 10 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
CAS No.: 76-61-9 Molecular Formula: C27H30O5S Formula Weight: 466.60 Physical Form: Crystalline Melting Point: 223° dec. Solubility: Insoluble in water. Soluble in alcohol, dilute alkali solutions MDL No.: MFCD00005869
pH indicator: 1.2 (red) to 2.8 (yellow) to 9.6 (blue)
Synonyme: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Nummer:76-61-9
- Formel:C₂₇H₃₀O₅S
- MDL-Nummer:MFCD00005869
- Molekulargewicht:466.6 g/mol
- Merck-Index:14,9400
- Schmelzender Platingehalt:223 °C (Zers.)
- Schmelzender Platingehalt:223 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:200-973-3
Dokumente
Zertifikate
Suche nach Chargennummerexternalurl.pdf msds_ALFA016272 sds_ALFA016272
Sicherheitsdatenblatt (SDS)
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:10 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



