Copper(II) gluconate
1 kg
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
CAS No.: 527-09-3 Molecular Formula: [HOCH2[CH(OH)]4CO2]2Cu Formula Weight: 453.85 Physical Form: Powder Melting Point: 155-157° MDL No.: MFCD00075297
Synonyme: Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate
- CAS Nummer:527-09-3
- Formel:C₁₂H₂₂CuO₁₄
- MDL-Nummer:MFCD00075297
- Molekulargewicht:453.84 g/mol
- Merck-Index:14,2640
- Schmelzender Platingehalt:155…157 °C
- Lagertemperatur:Raumtemperatur
- EINECS:208-408-2
Dokumente
Technische Daten
Technische Daten
- VE:1 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



