AuftragsverfolgungKontakt
Copper(II) gluconate
50 g
  15325.18
Copper(II) gluconate
  15325.18
 :  527-09-3
Copper(II) gluconate
Copper(II) gluconate
50 g
 :  527-09-3
SFr. 72.30
1 * 50 g
CAS No.: 527-09-3 Molecular Formula: [HOCH2[CH(OH)]4CO2]2Cu Formula Weight: 453.85 Physical Form: Powder Melting Point: 155-157° MDL No.: MFCD00075297
 :  Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate
  •  :527-09-3
  •  :C₁₂H₂₂CuO₁₄
  •  :MFCD00075297
  •  :453.84 g/mol
  •  :14,2640
  •  :155…157 °C
  •  :Raumtemperatur
  •  :208-408-2
  • VE:
    50 g