Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Natriumperiodat ≥98%

Natriumperiodat ≥98%

Glass bottle for solids, 500 g
Catalog # 13798.36
Oxidizing
Irritant
Natriumperiodat ≥98%
Catalog # 13798.36
CAS Number:  7790-28-5
Natriumperiodat ≥98%
Natriumperiodat ≥98%

Glass bottle for solids, 500 g

CAS Number:  7790-28-5
Price
538.00 SFr.
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 7790-28-5 Molecular Formula: INaO4 Formula Weight: 213.89 Physical Form: Powder Density: 3.86 Appearance: White Odour: Odourless Melting Point: 300° dec. Solubility: Soluble in water and acids Merck Reference: 14,8640 UN No.: 1479 MDL No.: MFCD00003534

Source of periodic acid, as an analytical reagent, and as an oxidizing agent

Synonyms:  Sodium metaperiodate
  • CAS Number:7790-28-5
  • Formula:NaIO₄
  • MDL Number:MFCD00003534
  • Molecular Weight:213.89 g/mol
  • Merck Index:14,8640
  • Boiling Pt:300 °C (1013 hPa)
  • Melting Pt:300 °C (Zers.)
  • Melting Pt:300 °C (Zersetzung)
  • Density:3,86
  • Storage Temperature:Raumtemperatur
  • EINECS:232-197-6
  • UN:1479
  • ADR:5.1,II
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.