Bariumfluorid ≥99%
Glass bottle for solids, 250 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 7787-32-8 Molecular Formula: BaF2 Formula Weight: 175.36 Physical Form: -40 Mesh Powder Density: 4.83 Refractive Index: 1.4741 Odour: Odourless Melting Point: 1353° Boiling Point: 2260° Solubility: Soluble in HCl, HNO3, HF, and aqueous solutions of ammonium chloride Merck Reference: 14,974 UN No.: 1564 MDL No.: MFCD00003450
Ceramic flux, glass manufacture, electronics, and dry-film lubricants
- CAS Nummer:7787-32-8
- Formel:BaF₂
- MDL-Nummer:MFCD00003450
- Molekulargewicht:175.33 g/mol
- Merck-Index:14,974
- Siedepunkt:2260 °C
- Schmelzender Platingehalt:1353 °C
- Dichte:4,83
- Lagertemperatur:Raumtemperatur
- EINECS:232-108-0
- UN:1564
- ADR:6.1,III
Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:250 g



