Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Bariumfluorid ≥99% | Avantor

Bariumfluorid ≥99%

Glass bottle for solids, 25 g
Bariumfluorid ≥99%
Katalognr. 12338.14
CAS Nummer:  7787-32-8
Bariumfluorid ≥99%
Bariumfluorid ≥99%

Glass bottle for solids, 25 g

CAS Nummer:  7787-32-8
Preis
SFr. 41.20
Einloggen für Verfügbarkeit
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 7787-32-8 Molecular Formula: BaF2 Formula Weight: 175.36 Physical Form: -40 Mesh Powder Density: 4.83 Refractive Index: 1.4741 Odour: Odourless Melting Point: 1353° Boiling Point: 2260° Solubility: Soluble in HCl, HNO3, HF, and aqueous solutions of ammonium chloride Merck Reference: 14,974 UN No.: 1564 MDL No.: MFCD00003450

Ceramic flux, glass manufacture, electronics, and dry-film lubricants

  • CAS Nummer:7787-32-8
  • Formel:BaF₂
  • MDL-Nummer:MFCD00003450
  • Molekulargewicht:175.33 g/mol
  • Merck-Index:14,974
  • Siedepunkt:2260 °C
  • Schmelzender Platingehalt:1353 °C
  • Dichte:4,83
  • Lagertemperatur:Raumtemperatur
  • EINECS:232-108-0
  • UN:1564
  • ADR:6.1,III

Dokumente

Technische Daten

Technische Daten

  • Packung:
    Glass bottle for solids
  • VE:
    25 g

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.