Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Kupfer(II)oxid ≥97%

Kupfer(II)oxid ≥97%

10 kg
Catalog # 12299.A9
Irritant
Environment
Kupfer(II)oxid ≥97%
Catalog # 12299.A9
CAS Number:  1317-38-0
Kupfer(II)oxid ≥97%
Kupfer(II)oxid ≥97%

10 kg

CAS Number:  1317-38-0
Price
SFr. 1’670.00
Log in for availability
1 * 10 kg
Product Details & Documents

Purity: 97% CAS No.: 1317-38-0 Molecular Formula: CuO Formula Weight: 79.55 Physical Form: -325 Mesh Powder Density: 6.3 Refractive Index: 2.63 Appearance: Black Odour: Odourless Melting Point: 1326° Solubility: Insoluble in water and alcohol. Soluble in dil. acids, NH4Cl and KCN Merck Reference: 14,2646 UN No.: 3077 MDL No.: MFCD00010979

As a pigment, in sweeting petroleum gases, in galvanic electrodes, metallurgical flux, as an optical glass polishing agent, and as a catalyst in organic reactions

Synonyms:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Number:1317-38-0
  • Formula:CuO
  • MDL Number:MFCD00010979
  • Molecular Weight:79.55 g/mol
  • Merck Index:14,2646
  • Boiling Pt:2000 °C (1013 hPa)
  • Melting Pt:1326 °C
  • Density:6,3
  • Storage Temperature:Raumtemperatur
  • EINECS:215-269-1
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • VE:
    10 kg

Specifications