Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Kupfer(II)oxid ≥97% | Avantor

Kupfer(II)oxid ≥97%

2 kg
Katalognr. 12299.A3
Reizend
Umfeld
Kupfer(II)oxid ≥97%
Katalognr. 12299.A3
CAS Nummer:  1317-38-0
Kupfer(II)oxid ≥97%
Kupfer(II)oxid ≥97%

2 kg

CAS Nummer:  1317-38-0
Preis
SFr. 447.00
Einloggen für Verfügbarkeit
1 * 2 kg
Product Details & Documents

Purity: 97% CAS No.: 1317-38-0 Molecular Formula: CuO Formula Weight: 79.55 Physical Form: -325 Mesh Powder Density: 6.3 Refractive Index: 2.63 Appearance: Black Odour: Odourless Melting Point: 1326° Solubility: Insoluble in water and alcohol. Soluble in dil. acids, NH4Cl and KCN Merck Reference: 14,2646 UN No.: 3077 MDL No.: MFCD00010979

As a pigment, in sweeting petroleum gases, in galvanic electrodes, metallurgical flux, as an optical glass polishing agent, and as a catalyst in organic reactions

Synonyme:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Nummer:1317-38-0
  • Formel:CuO
  • MDL-Nummer:MFCD00010979
  • Molekulargewicht:79.55 g/mol
  • Merck-Index:14,2646
  • Siedepunkt:2000 °C (1013 hPa)
  • Schmelzender Platingehalt:1326 °C
  • Dichte:6,3
  • Lagertemperatur:Raumtemperatur
  • EINECS:215-269-1
  • UN:3077
  • ADR:9,III

Dokumente

Technische Daten

Technische Daten

  • VE:
    2 kg

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.