Calciumfluorid ≥99%
Glass bottle for solids, 10 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Physical Form: Transparent Crystals Density: 3.18 Refractive Index: 1.434 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 1403° Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907
- CAS Nummer:7789-75-5
- Formel:CaF₂
- MDL-Nummer:MFCD00010907
- Molekulargewicht:78.07 g/mol
- Merck-Index:14,1667
- Siedepunkt:∼2500 °C
- Schmelzender Platingehalt:1403 °C
- Dichte:3,18
- Lagertemperatur:Raumtemperatur
- EINECS:232-188-7
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:10 g



