Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

n-Hexan ≥95,0%, BAKER ANALYZED® HPLC, J.T.Baker®

Glasflasche, 2,5 L
Catalog # 9304.2500
Supplier:   Avantor
n-Hexan ≥95,0%, BAKER ANALYZED® HPLC, J.T.Baker®
Catalog # 9304.2500
Supplier:  Avantor
CAS Number:  110-54-3
n-Hexan ≥95,0%, BAKER ANALYZED® HPLC, J.T.Baker®
n-Hexan ≥95,0%, BAKER ANALYZED® HPLC, J.T.Baker®

Glasflasche, 2,5 L

CAS Number:  110-54-3
Price
SFr. 325.00
Log in for availability
1 * 2,5 L
Product Details & Documents

for use in High Performance Liquid Chromatography

Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • UN:1208
  • ADR:3,II
Specifications

Specification Test Results

  • Assay (by GC):
    min. 95 %
  • Residue after Evaporation:
    max. 2 ppm
  • Substances Darkened by H2SO4:
    passes test
  • Water (H2O):
    max. 0,01 %
  • Ultraviolet Absorbance,(1.00-cm path vs water; curve smooth throughout stated range with no extraneous impurity peaks).:
    -
  • at 210 nm:
    max. 0,2
  • at 220 nm:
    max. 0,08
  • at 254 nm:
    max. 0,008
  • at 280 nm:
    max. 0,005
  • at 350 nm:
    max. 0,005
  • UV Cut-off, nm:
    max. 192

Specifications

  • Packung:
    Glasflasche
  • VE:
    2,5 L

Specifications