Receiving flasks
2000 ml, P+G coated, KS 35/20
Katalognr. 531-1846
Anbieter: BUCHI
Direkt zu
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
Technische Daten
- Kugelschliff-Schale:KS 35/20
- Typ:P+G coated
- Kapazität:2000 ml