Jump to:
Product Details & Documents
Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
- CAS Number:643-79-8
- Formula:C₈H₆O₂
- MDL Number:MFCD00003335
- Molecular Weight:134.13 g/mol
- Boiling Pt:266 °C (1013 hPa)
- Melting Pt:54…56 °C
- Density:1,13 g/cm³ (20 °C)
- Flash Pt:132 °C
- Storage Temperature:Kühlschrank
- EINECS:211-402-2
- ADR:8 (6.1),
- :M-Clarity™ Qualitätsstufe = MQ100
- UN:2923
Documents
Specifications
Specifications
- Packung:Glasflasche
- VE:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



