Tweezers
180 mm
Katalognr. 232-1951
Anbieter: VITLAB
Direkt zu
Product Details & Documents
POM, yellow. Universal tweezers for a variety of applications.
- Blunt, elastic, very good resilience
- With grooves on the outside for optimum handling and grip
- Good chemical resistance
Technische Daten
- Länge:180 mm