Stativringe, schwer
Schwerer Stativring, mit Muffe, 50 mm
Catalog # 241-0320
Supplier: RSG
Jump to:
Product Details & Documents
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
Specifications
- Ø int.:50 mm
- Bezeichnung:Schwerer Stativring, mit Muffe