Pierce™ Disposable Columns
Disposable PS columns, for packing 1 - 5 ml bed volumes
Catalog # PIER29922
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Empty polystyrene columns with matching frits and caps for use in a variety of resin based affinity or desalting techniques.
- For benchtop gravity flow affinity purification and size exclusion chromatography
- Supplied complete with porous polyethylene discs, stoppers and end caps
- Compatible with most types of aqueous buffer eluents
- Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Specifications
- Bezeichnung:Disposable PS columns, for packing 1 - 5 ml bed volumes
- Seq:02
Accessories
Product Information
- Catalog #BezeichnungAvailabilityPrice
- PIER29923Einweg-PP-Trichter (Pufferspeicher) der auf Pierce™-Einwegsäulen passt.Log in for availability1 * 50 ST - SFr. 304.00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.