Öle für Rotationspumpen
Product Details & Documents
Rotary vane pump oil, B
Rotary vane pump oil, B has excellent viscosity characteristics and low vapour pressure. Its exceptional chemical resistance, more stable behaviour when pumping oxidants, acid and alkaline vapours, and the fact that it is better tolerated on contact with skin, sets this oil apart from conventional mineral oils. Even after continuous operation for several weeks and months, the oil ensures the pressure is maintained constantly. Used for priming all RE/RZ series pumps.
Rotationspumpen-Öl, K8Das Rotationspumpenöl K 8 ist ein Spezialöl, das für das Absaugen von Säuredämpfen vorgesehen ist. Dieses Öl ist stark hygroskopisch und deshalb nur bedingt zum Pumpen von Wasserdampf geeignet. Der in diesem Öl enthaltene alkalische Korrosionsschutz verbraucht sich im Laufe des Betriebs. Es ist deshalb notwendig, je nach Art und Menge der Dämpfe dieses Öl regelmäßig zu wechseln. Vor dem Stilllegen der Pumpe über mehrere Tage oder länger muss das Öl gewechselt werden.
Silicone oil, SI 2
Silicone oil, SI 2 is designed to be used when extremely high durability is required. Because of its chemical composition, the oil is not affected by certain aggressive gases (particularly chlorine and hydrogen chloride) or solvent-containing vapours. It is also resistant to most acids and acid solutions.
Low vapour pressure even at high temperatures
Outstanding lubricating properties
Low oil backflow
Good durability, resistance to cracking
Good resistance to oxidation
Silicone oil, SI 2 is designed to be used when extremely high durability is required. Because of its chemical composition, the oil is not affected by certain aggressive gases (particularly chlorine and hydrogen chloride) or solvent-containing vapours. It is also resistant to most acids and acid solutions.
Technische Daten
- Bezeichnung:Öl für Drehschieberpumpen, B
- Seq:02