Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Power supply adapters

Adapter set, 2 mm male/ 4 mm female, for electrophoresis systems
  18-1129-59
 :   Cytiva
Power supply adapters
  18-1129-59
 :  Cytiva
Power supply adapters

Adapter set, 2 mm male/ 4 mm female, for electrophoresis systems

 
SFr. 97.90
   
1 * 1 SET
 

Choice of adapters for connecting 4 or 2 mm leads to 2 or 4 mm output jacks, respectively.

  • Approved for 500 V relative protection ground
  • Adapter set 2 mm M/4 mm F for connecting sheathed 2 mm male plug instrument leads to 4 mm sheathed output jacks
  • Adapter set 4 mm M/2 mm F for connecting sheathed 4 mm male plug instrument leads to 2 mm high voltage output jacks
 
  • Bezeichnung:
    Adapter set, 2 mm male/ 4 mm female, for electrophoresis systems