Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Power supply adapters | Avantor

Power supply adapters

Adapter set, 4 mm male/ 2 mm female, for electrophoresis systems
Katalognr. 18-1129-58
Anbieter:   Cytiva
Power supply adapters
Katalognr. 18-1129-58
Anbieter:  Cytiva
Power supply adapters

Adapter set, 4 mm male/ 2 mm female, for electrophoresis systems

Preis
SFr. 144.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Choice of adapters for connecting 4 or 2 mm leads to 2 or 4 mm output jacks, respectively.

  • Approved for 500 V relative protection ground
  • Adapter set 2 mm M/4 mm F for connecting sheathed 2 mm male plug instrument leads to 4 mm sheathed output jacks
  • Adapter set 4 mm M/2 mm F for connecting sheathed 4 mm male plug instrument leads to 2 mm high voltage output jacks
Technische Daten
  • Bezeichnung:
    Adapter set, 4 mm male/ 2 mm female, for electrophoresis systems
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.