Instrument trays
260×360×50 mm
Katalognr. USBE4743
Anbieter: USBECK Laborgeräte
Direkt zu
Product Details & Documents
Made of stainless steel.
- Rounded corners
- With rim
- Very good chemical and temperature resistance
Technische Daten
- B×T×H:260×360×50 mm