Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Burner, Teclu

Burner, Teclu

Teclu burner with air regulation, propane gas, tubing Ø 14 mm, Propan, 17 mm
Catalog # USBE1201
Supplier:   USBECK Laborgeräte
Burner, Teclu
Catalog # USBE1201
Burner, Teclu
Burner, Teclu

Teclu burner with air regulation, propane gas, tubing Ø 14 mm, Propan, 17 mm

Price
SFr. 62.40
Log in for availability
1 * 1 ST
Product Details & Documents

Nickel plated brass base, in powder coated grey.

  • With air regulation valve and throttle screw
  • Without pilot light

Height: 165 mm

Specifications
  • Bezeichnung:
    Teclu burner with air regulation, propane gas, tubing Ø 14 mm
  • Ø Kopf:
    17 mm
  • Für:
    Propan
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.