AuftragsverfolgungKontakt
Dip slides
Dip slides
PCA / Ogy agar, 10
Codice catalogo OXOIDS0170A
Nome del fornitore:   THERMO FISHER DIAGNOSTICS
Dip slides
Codice catalogo OXOIDS0170A
Nome del fornitore:  THERMO FISHER DIAGNOSTICS
Numero CAS:  
Dip slides
PCA / Ogy agar, 10
Numero CAS:  
Price
SFr. 48.90
Log in per disponibilità
1 * 10 ST
Product Details & Documents
Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor’s surgery and then returned to its plastic container for easy transport back to the laboratory.
  • Make colony counting for the bacterial assessment simple
Documents
Specifiche
Specifiche
  • VE:
    10
  • Bezeichnung:
    PCA / Ogy agar
Specifiche