AuftragsverfolgungKontakt
Dip slides | Avantor
Dip slides
TTC Red Spot, 10
Katalognr. OXOIDS0147A
Dip slides
Katalognr. OXOIDS0147A
CAS Nummer:  
Dip slides
TTC Red Spot, 10
CAS Nummer:  
Preis
SFr. 49.40
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor’s surgery and then returned to its plastic container for easy transport back to the laboratory.
  • Make colony counting for the bacterial assessment simple
Dokumente
Technische Daten
Technische Daten
  • VE:
    10
  • Bezeichnung:
    TTC Red Spot