Paraldehyd
Glasflasche, 1 L
Anbieter: Merck
Direkt zu
Product Details & Documents
- CAS Nummer:123-63-7
- Formel:C₆H₁₂O₃
- MDL-Nummer:MFCD00036208
- Molekulargewicht:132.16 g/mol
- Lagertemperatur:Raumtemperatur
- ADR:3,III
- :M-Clarity™ Qualitätsstufe = MQ200
- EINECS:204-639-8
- UN:1264
Dokumente
Technische Daten
Ergebnisse des Spezifikationstests
- Solubility:120 g/l (13 °C)
- Melting Point:12 °C
- Molar Mass:132.15 g/mol
- Boiling Point:124 °C (1013 hPa)
- Vapor Pressure:13.3 hPa (20 °C)
- Flash Point:27 °C
- Refractive Index:1.4049 (20 °C)
- Explosion Limit:1.3 - 17.0 %(V)
- Density:0.99 g/cm3 (20 °C)
- Ignition Point:235 °C
Technische Daten
- Packung:Glasflasche
- VE:1 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



