Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Cell culture flasks, Nunc™ TripleFlask™ | Avantor

Cell culture flasks, Nunc™ TripleFlask™

500 cm², Belüftet/geschlossen, Gerade, Nunclon™Δ coated, 100 - 200 ml, 1/pack
Katalognr. 734-2000
Cell culture flasks, Nunc™ TripleFlask™
Katalognr. 734-2000
Cell culture flasks, Nunc™ TripleFlask™
Cell culture flasks, Nunc™ TripleFlask™

500 cm², Belüftet/geschlossen, Gerade, Nunclon™Δ coated, 100 - 200 ml, 1/pack

Preis
SFr. 928.00
Einloggen für Verfügbarkeit
1 * 32 ST
Product Details & Documents

The Nunc™ TripleFlask™ is a 3-layer flask providing three times the growth surface of a T175 flask for the same footprint, saving space in the incubator.

  • Recommended working volume is 200 ml
  • Promotes monolayer adherent cell culture
  • Also available barcoded (*) with large Code 128
  • Nunclon Delta Certificate is included in each case for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
  • Available in non-treated format

Perform cell culture expansion and experience 500 cm² total growth area within the footprint of a standard flask using Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems.

Nunc™ Delta surface treatment offers maximum adhesion for a broad range of cell types. Barcoding option for automation cell culture.

Technische Daten
  • Verschlussart:
    Belüftet/geschlossen
  • Wachstumsfläche:
    500 cm²
  • Verpackt:
    1/pack
  • Empf. Arbeitsvolumen:
    100 - 200 ml
  • Ausführung:
    Nunclon™Δ coated
  • Seq:
    03
  • Hals:
    Gerade