Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ Vector Kit, 20 Tests
Katalognr. 70595-3
pSTBlue-1 AccepTor™ vector kit
Katalognr. 70595-3
CAS Nummer:  
pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ Vector Kit, 20 Tests
CAS Nummer:  
Preis
SFr. 483.00
Einloggen für Verfügbarkeit
1 * 20 Tests
Product Details & Documents
The pSTBlue-1 AccepTor™ vector kits are designed to simplify the PCR cloning process by using non-proofreading thermostable DNA polymerases (eg. KOD XL polymerase and native and recombinant Taq polymerases), which leave single 3'-dA overhangs on the reaction products. The AccepTor™ vectors enable direct ligation by virtue of single 3'-dU overhangs that anneal efficiently with 3'-dA extensions on PCR products. The dU residues are converted to dT residues in vivo following transformation.
Informazioni per l'ordine: AccepTor™ vectors are supplied in a ready-to-ligate format.
Technische Daten
Technische Daten
  • VE:
    20 Tests
  • Bezeichnung:
    pSTBlue-1 AccepTor™ Vector Kit
Technische Daten
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..