Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
FMOC-D-THR(TBU)-OH 1 * 1 g

FMOC-D-THR(TBU)-OH 1 * 1 g

Catalog # 04-13-1053-1
FMOC-D-THR(TBU)-OH 1 * 1 g
Catalog # 04-13-1053-1
Supplier:  
CAS Number:  
FMOC-D-THR(TBU)-OH 1 * 1 g
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FMOC-D-THR(TBU)-OH 1 * 5 g Catalog #04-13-1053-5

Product 04-13-1053-5 is a replacement for 04-13-1053-1 but varies by Brand, Packaging or Quantity.

Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.