Micro control valves
Manual on/off valve, pneumatic (MOVP), "L" single outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
Katalognr. SGEA1236091
Anbieter: Trajan Scientific and Medical
Direkt zu
Product Details & Documents
Micro control valves provide precision metering and on-off control of gas flows. They are designed specifically for chromatography applications and used at elevated temperatures from high vacuum to medium pressure.
Actuated remotely with a toggle switch or solenoid valve, and a 50 to 55 psi air supply.
Technische Daten
- Bezeichnung:Manual on/off valve, pneumatic (MOVP), "L" single outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
Accessories
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis