Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
BOC-ASP-OH 1 * 5 g

BOC-ASP-OH 1 * 5 g

Catalog # 04-12-0129-5
BOC-ASP-OH 1 * 5 g
Catalog # 04-12-0129-5
Supplier:  
CAS Number:  
BOC-ASP-OH 1 * 5 g
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BOC-ASP-OH 1 * 25 g Catalog #04-12-0129-25

Product 04-12-0129-25 is a replacement for 04-12-0129-5 but varies by Brand, Packaging or Quantity.

Product Details & Documents