Natriumpyruvat, Millipore®
Kunststoffflasche, 250 g
Anbieter: Merck
Direkt zu
Product Details & Documents
Synonyme: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Nummer:113-24-6
- Formel:CH₃COCOONa
- MDL-Nummer:MFCD00002586
- Molekulargewicht:110.04 g/mol
- Schmelzender Platingehalt:220…230 °C
- Dichte:1,718 g/cm³ (20 °C)
- :M-Clarity™ Qualitätsstufe = MQ100
- Lagertemperatur:Raumtemperatur
- EINECS:204-024-4
Technische Daten
Technische Daten
- Packung:Kunststoffflasche
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



