Jump to:
Product Details & Documents
Synonyms: Copper aluminium zinc alloy (50:45:5 w%), Devarda's metal
- CAS Number:8049-11-4
- MDL Number:MFCD00198176
- Merck Index:13,02958
- Boiling Pt:906 °C (1013 hPa)
- Melting Pt:490…560 °C
- Density:5,79 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- UN:3089
- ADR:4.1,II
- :M-Clarity™ Qualitätsstufe = MQ300
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:490 - 560 °C
- Bulk Density:2080 kg/m3
- Boiling Point:906 °C
- Density:5.79 g/cm3 (20 °C)
Specifications
- Packung:Kunststoffflasche
- VE:1 kg



