2-Oxoglutarsäure
Kunststoffflasche, 1 kg
Anbieter: Merck
Direkt zu
Product Details & Documents
Synonyme: 2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid
- CAS Nummer:328-50-7
- Formel:HOOCCH₂CH₂COCOOH
- MDL-Nummer:MFCD00004165
- Molekulargewicht:146.1 g/mol
- Merck-Index:13,05320
- Siedepunkt:323 °C (1013 hPa)
- Schmelzender Platingehalt:112…116 °C
- :M-Clarity™ Qualitätsstufe = MQ100
- Lagertemperatur:Kühlschrank
- EINECS:206-330-3
Documents
Technische Daten
Technische Daten
- Packung:Kunststoffflasche
- VE:1 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



