Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
n-Hexan, EMPLURA®, Supelco®

n-Hexan, EMPLURA®, Supelco®

Glasflasche, 2,5 L
Catalog # 1.04368.2500
Flammable
Health
Irritant
Environment
Supplier:   Merck
n-Hexan, EMPLURA®, Supelco®
Catalog # 1.04368.2500
Supplier:  Merck
CAS Number:  110-54-3
n-Hexan, EMPLURA®, Supelco®
n-Hexan, EMPLURA®, Supelco®

Glasflasche, 2,5 L

CAS Number:  110-54-3
Price
SFr. 178.00
Log in for availability
1 * 2,5 L
Product Details & Documents

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Raumtemperatur
  • UN:1208
  • ADR:3,II
  •  :M-Clarity™ Qualitätsstufe = MQ200
  • EINECS:203-777-6

Documents

Specifications

Specifications

  • Packung:
    Glasflasche
  • VE:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.