n-Hexan, EMPLURA®, Supelco®
Glasflasche, 1 L
Supplier: Merck
Jump to:
Product Details & Documents
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–94,3 °C
- Density:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Raumtemperatur
- UN:1208
- ADR:3,II
- :M-Clarity™ Qualitätsstufe = MQ200
- EINECS:203-777-6
Documents
Certificates
Search by Lot Numberexternalurl.pdf ProductPage_104368
Product specification
msds_104368
SDS
Specifications
Specifications
- Packung:Glasflasche
- VE:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



