Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

L(+)-Histidin, Millipore®

Kunststofftrommel, 10 kg
Catalog # 1.04351.9010
Supplier:   Merck
L(+)-Histidin, Millipore®
Catalog # 1.04351.9010
Supplier:  Merck
CAS Number:  71-00-1
L(+)-Histidin, Millipore®
L(+)-Histidin, Millipore®

Kunststofftrommel, 10 kg

CAS Number:  71-00-1
Price
Inquire For Price
Log in for availability
1 * 10 kg
Product Details & Documents

Soluble in water and mineral acids; insoluble in organic solvents.

Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1,44 g/cm³ (20 °C)
  •  :M-Clarity™ Qualitätsstufe = MQ100
  • Storage Temperature:Raumtemperatur
  • EINECS:200-745-3
Specifications

Specifications

  • Packung:
    Kunststofftrommel
  • VE:
    10 kg

Specifications