Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
L(+)-Histidin, Millipore®

L(+)-Histidin, Millipore®

Glasflasche, 1 kg
Catalog # 1.04351.1000
Supplier:   Merck
L(+)-Histidin, Millipore®
Catalog # 1.04351.1000
Supplier:  Merck
CAS Number:  71-00-1
L(+)-Histidin, Millipore®
L(+)-Histidin, Millipore®

Glasflasche, 1 kg

CAS Number:  71-00-1
Price
SFr. 1’110.00
Log in for availability
1 * 1 kg
Product Details & Documents

Soluble in water and mineral acids; insoluble in organic solvents.

Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1,44 g/cm³ (20 °C)
  •  :M-Clarity™ Qualitätsstufe = MQ100
  • Storage Temperature:Raumtemperatur
  • EINECS:200-745-3
Specifications

Specifications

  • Packung:
    Glasflasche
  • VE:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.