Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
L(+)-Cystein, Millipore®

L(+)-Cystein, Millipore®

Kunststoffflasche, 25 g
Catalog # 1.02838.0025
Irritant
Supplier:   Merck
L(+)-Cystein, Millipore®
Catalog # 1.02838.0025
Supplier:  Merck
CAS Number:  52-90-4
L(+)-Cystein, Millipore®

Kunststoffflasche, 25 g

CAS Number:  52-90-4
Price
67.80 SFr.
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS Number:52-90-4
  • Formula:HSCH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Merck Index:13,02810
  • Boiling Pt:254 °C (1013 hPa)
  • Melting Pt:115 °C
  • Density:1,523 g/cm³ (20 °C)
  •  :M-Clarity™ Qualitätsstufe = MQ100
  • Storage Temperature:Raumtemperatur
  • EINECS:200-158-2
Specifications

Specifications

  • Packung:
    Kunststoffflasche
  • VE:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.