Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
D(+)-Cellobiose, Millipore®

D(+)-Cellobiose, Millipore®

Glasflasche, 25 g
Catalog # 1.02352.0025
Supplier:   Merck
D(+)-Cellobiose, Millipore®
Catalog # 1.02352.0025
Supplier:  Merck
CAS Number:  528-50-7
D(+)-Cellobiose, Millipore®

Glasflasche, 25 g

CAS Number:  528-50-7
Price
SFr. 296.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
  • CAS Number:528-50-7
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00136034
  • Molecular Weight:342.30 g/mol
  •  :M-Clarity™ Qualitätsstufe = MQ100
  • Storage Temperature:Raumtemperatur
  • EINECS:208-436-5

Documents

Specifications

Specifications

  • Packung:
    Glasflasche
  • VE:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.