Jump to:
Product Details & Documents
- CAS Number:534-17-8
- Formula:Cs₂CO₃
- MDL Number:MFCD00010957
- Molecular Weight:325.82 g/mol
- Merck Index:13,02020
- Melting Pt:610 °C
- Density:4,24 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:208-591-9
Documents
externalurl.pdf ProductPage_102040
Product specification
msds_102040
SDS
Specifications
Specifications
- Packung:Glasflasche
- VE:25 g



