Kryoschreiber-Set, Nalgene®
Kryo-Schreiber-Set, Schwarz
For permanent marking of cardboard, cloth labels and plastic.
- Ink will not fade at ultra-low temperatures
- Extra fine point
Set consists of 1 red, 1 green, 1 blue and 1 black marker.
- Farbe:Schwarz
- Bezeichnung:Kryo-Schreiber-Set
- Cat. No.:479-0074
- Umweltzertifizierung:NAR Certificate of Retirement
- Umweltfreundlichkeit:Umweltschonende Herstellung