Acrylamid
Glass bottle for solids, 500 g
Katalognr. NADIEC-201-500G
Marketsource
Anbieter: NATIONAL DIAGNOSTICS
Direkt zu
Product Details & Documents
Synonyme: Prop-2-enamide
- CAS Nummer:79-06-1
- Formel:C₃H₅NO
- MDL-Nummer:MFCD00008032
- Molekulargewicht:71.08 g/mol
- Siedepunkt:232 °C (1013 hPa)
- Schmelzender Platingehalt:84,5 °C
- Dichte:1,122 g/cm³ (20 °C)
- Blitz Pt:138 °C
- Lagertemperatur:Kühlschrank
- EINECS:201-173-7
- UN:2074
- ADR:6.1,III
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:500 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



