Objectives for DMW-B1
Plan phase objective (spring), 40×/0,65
Katalognr. 632-8994
Anbieter: MOTIC
Direkt zu
Product Details & Documents
Plan phase objectives for use with the digital microscope DMW-B1.
Technische Daten
- Vergrößerung:40×/0,65
- Bezeichnung:Plan phase objective (spring)