AuftragsverfolgungKontakt
Extractor heads for specific light solvents
250 ml
  LENZ05432149
Extractor heads for specific light solvents
  LENZ05432149
Extractor heads for specific light solvents
Extractor heads for specific light solvents
250 ml
SFr. 444.00
1 * 1 ST
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific light solvents, with insert filter (porosity: 0).
  • Extraktorinhalt:
    250 ml
  • Seq:
    02