Kühler, Davies
250 mm, 24/29 NS, 24/29 NS
Katalognr. LENZ05215303
Anbieter: Lenz Laborglas
Direkt zu
Product Details & Documents
DURAN® borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two glass olives.
Dokumente
BSE TSE Glass Columns
BSE/TSE-Zertifikate
Technische Daten
- Kern:24/29 NS
- Mantellänge:250 mm
- Hülse:24/29 NS