Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Kühler, Davies | Avantor
Kühler, Davies
250 mm, 24/29 NS, 24/29 NS
Katalognr. LENZ05215303
Anbieter:   Lenz Laborglas
Kühler, Davies
Katalognr. LENZ05215303
Anbieter:  Lenz Laborglas
Kühler, Davies
Kühler, Davies
250 mm, 24/29 NS, 24/29 NS
Preis
SFr. 195.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two glass olives.
Dokumente
Technische Daten
  • Kern:
    24/29 NS
  • Mantellänge:
    250 mm
  • Hülse:
    24/29 NS
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.